Dimethyl acetal, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Acetaldehyde dimethyl acetal, 1,1-Dimethoxyethane
M-Clarity™ quality level = MQ300
W342602-100G W342602-1KG W342602-SAMPLE
SIALW342602-100GEA 121 EUR
SIALW342602-100G SIALW342602-1KG SIALW342602-SAMPLE
Dimethyl acetal, Sigma-Aldrich®
Dimethyl acetal
Formula: C₄H₁₀O₂
MW: 90.12 g/mol
Boiling Pt: 64
Melting Pt: –113 °C
Density: 0.8505 g/cm³ (20 °C)
MDL Number: MFCD00008493
CAS Number: 534-15-6
EINECS: 208-589-8
UN: 2377
ADR: 3 II
Merck Index: 13,03253