Valeric anhydride, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ100
245933-100ML 245933-500ML
SIAL245933-100MLEA 125 EUR
SIAL245933-100ML SIAL245933-500ML
Valeric anhydride, Sigma-Aldrich®
Valeric anhydride
Formula: C₁₀H₁₈O₃
Boiling Pt: 228 °C (1013 hPa)
Melting Pt: –56 °C
Density: 0.94 g/cm³ (20 °C)
MDL Number: MFCD00009477
CAS Number: 2082-59-9
EINECS: 218-212-9
UN: 3265
ADR: 8,III