2-Propanesulphonyl chloride, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Isopropylsulphonyl chloride
M-Clarity™ quality level = MQ100
242705-100G 242705-25G 242705-5G
SIAL242705-100GEA 295 EUR
SIAL242705-100G SIAL242705-25G SIAL242705-5G
2-Propanesulphonyl chloride, Sigma-Aldrich®
2-Propanesulphonyl chloride
Formula: C₃H₇ClO₂S
Boiling Pt: 181 °C (1013 hPa)
Melting Pt: –47 °C
Density: 1.273 g/cm³ (20 °C)
Flash Pt: 86 °C
MDL Number: MFCD00007453
CAS Number: 10147-37-2
EINECS: 233-415-2
UN: 3265
ADR: 8,II