1-Pentanesulphonic acid sodium salt, Sigma-Aldrich®
Supplier: Merck
Synonyms: Sodium 1-pentanesulphonate, Sodium pentane-1-sulphonate, Pentane-1-sulphonic acid sodium salt, Sodium pentanesulphonate, 98%, Sodium pentanesulphonate
M-Clarity™ quality level = MQ100
P0299-100G P0299-25G
SIALP0299-100GEA 678 EUR
SIALP0299-100G SIALP0299-25G
1-Pentanesulphonic acid sodium salt, Sigma-Aldrich®
1-Pentanesulphonic acid sodium salt
Formula: C₅H₁₁O₃SNa
MW: 174.2 g/mol
Melting Pt: > 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00007541
CAS Number: 22767-49-3
EINECS: 245-208-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ddb1c23c8a6fe3c156a3aacb62721e1d1a34efb0&vid=1936dcdb47ba374ce21fd758660f7ca6ba95d254&prid=web&clid=SAPDAM