Isoamyl alcohol, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Isopentyl alcohol, iso-Pentanol, 3-Methyl-1-butanol, 3-Methylbutanol
M-Clarity™ quality level = MQ300
W205702-1KG-K W205702-20KG-K W205702-8KG-K
SIALW205702-1KG-KEA 79.9 EUR
SIALW205702-1KG-K SIALW205702-20KG-K SIALW205702-8KG-K
Isoamyl alcohol, Sigma-Aldrich®
Isoamyl alcohol
Formula: (CH₃)₂CHCH₂CH₂OH
MW: 88.15 g/mol
Boiling Pt: 130
Melting Pt: –117 °C
Density: 0.812 g/cm³ (20 °C)
Flash Pt: 42
Storage Temperature: Ambient
MDL Number: MFCD00002934
CAS Number: 123-51-3
EINECS: 204-633-5
UN: 1105
ADR: 3 III
Merck Index: 13,05214
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cd7b89cc1890b5d4b2083195c30c0d1047c32e9a&vid=170c60e26c7b4edab6e0686f91d04e676da16e6d&prid=web&clid=SAPDAM