Pinacol, Sigma-Aldrich®
Supplier: Merck
Synonyms:
2,3-Dimethylbutane-2,3-diol, Tetramethylethylene glycol, 2,3-Dimethyl-2,3-butanediol
M-Clarity™ quality level = MQ200
221171-100G
221171-500G
221171-5G
SIAL221171-100GEA
154
EUR
SIAL221171-100G
SIAL221171-500G
SIAL221171-5G
Pinacol, Sigma-Aldrich®
Pinacol
Formula:
C₆H₁₄O₂ Boiling Pt: 174 °C (1013 hPa) Melting Pt: 40…41 °C Density: 0.967 g/cm³ (20 °C) Flash Pt: 77 °C |
MDL Number:
MFCD00004462 CAS Number: 76-09-5 EINECS: 200-933-5 Merck Index: 13,07521 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...