Cyclohexylacetic acid, Sigma-Aldrich®
Supplier: Merck
Synonyms: Cyclohexaneacetic acid
M-Clarity™ quality level = MQ100
C104507-25G C104507-5G
SIALC104507-25GEA 145 EUR
SIALC104507-25G SIALC104507-5G
Cyclohexylacetic acid, Sigma-Aldrich®
Cyclohexylacetic acid
Formula: C₈H₁₄O₂
MW: 142.2 g/mol
Boiling Pt: 242
Melting Pt: 26…27 °C
Density: 0.9985 g/cm³ (22 °C)
MDL Number: MFCD00001518
CAS Number: 5292-21-7
EINECS: 226-132-0