Allyl isothiocyanate, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
377430-100G 377430-500G 377430-5G
SIAL377430-100GEA 60.1 EUR
SIAL377430-100G SIAL377430-500G SIAL377430-5G
Allyl isothiocyanate, Sigma-Aldrich®
Allyl isothiocyanate
Formula: C₄H₅NS
Boiling Pt: 148 °C (1013 hPa)
Melting Pt: –80 °C
Density: 1.02 g/cm³ (20 °C)
Flash Pt: 46 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004822
CAS Number: 57-06-7
EINECS: 200-309-2
UN: 1545
ADR: 6.1,II
Merck Index: 13,00292