9-Anthracenecarboxylic acid, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
A89405-25G A89405-5G
SIALA89405-25GEA 222 EUR
SIALA89405-25G SIALA89405-5G
9-Anthracenecarboxylic acid, Sigma-Aldrich®
9-Anthracenecarboxylic acid
Formula: C₁₅H₁₀O₂
Boiling Pt: 411 °C (1013 hPa)
Melting Pt: 214…217 °C
Density: 1.397 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001257
CAS Number: 723-62-6
EINECS: 211-964-9