MES sodium salt, Sigma-Aldrich®
Supplier: Merck
Synonyms: Sodium 2-Morpholinoethanesulphonate, Sodium 3-morpholin-1-ylethylsulphonate, 2-(N-Morpholino)ethanesulphonic acid sodium salt, Sodium 2-morpholinoethane-1-sulphonate, MES-Na
M-Clarity™ quality level = MQ400
M3058-100G M3058-1KG M3058-25G M3058-500G
SIALM3058-100GEA 288 EUR
SIALM3058-100G SIALM3058-1KG SIALM3058-25G SIALM3058-500G
MES sodium salt, Sigma-Aldrich®
MES sodium salt
Formula: C₆H₁₂NNaO₄S
MW: 217.22 g/mol
MDL Number: MFCD00065473
CAS Number: 71119-23-8
EINECS: 275-203-2