Penicillin G sodium salt, Sigma-Aldrich®
Supplier: Merck
Synonyms: 6-(2-Phenylacetamido)penicillanic acid sodium salt, Benzylpenicillin sodium salt, Sodium [2S-(2α,5α,6β)]-3,3-dimethyl-7-oxo-6-(phenylacetamido)-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
MQ200
13752-1G-F 13752-5G-F
SIAL13752-1G-FEA 22.4 EUR
SIAL13752-1G-F SIAL13752-5G-F
Penicillin G sodium salt, Sigma-Aldrich®
Penicillin G sodium salt
Formula: C₁₆H₁₇N₂NaO₄S
MW: 356.38 g/mol
Melting Pt: 255
Density: 1.38 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00069666
CAS Number: 69-57-8
EINECS: 200-710-2