Benzamidine, Sigma-Aldrich®
Supplier: Merck
Warning
12072-50G 12072-10G
SIAL12072-50GEA 726 EUR
SIAL12072-50G SIAL12072-10G
Benzamidine, Sigma-Aldrich®
Benzamidine
Formula: C₇H₈N₂
MW: 120.15 g/mol
MDL Number: MFCD00042826
CAS Number: 618-39-3
EINECS: 210-546-3