Octyl-3,4,5-trihydroxybenzoate, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Octyl 3,4,5-trihydroxybenzoate, Gallic acid octyl ester, Octyl gallate, n-Octyl gallate
48700-250G-F 48700-50G-F
SIAL48700-250G-FEA 481 EUR
SIAL48700-250G-F SIAL48700-50G-F
Octyl-3,4,5-trihydroxybenzoate, Sigma-Aldrich®
Octyl 3,4,5-trihydroxybenzoate
Formula: C₁₅H₂₂O₅
Melting Pt: 95…100 °C
MDL Number: MFCD00002197
CAS Number: 1034-01-1
EINECS: 213-853-0