Propyl-3,4,5-trihydroxybenzoate, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Gallic acid propyl ester, Propyl gallate, Propyl 3,4,5-trihydroxybenzoate, Progalline P, N-Propyl gallate
M-Clarity™ quality level = MQ300
W294705-10KG-K W294705-1KG-K
SIALW294705-10KG-KEA 958 EUR
SIALW294705-10KG-K SIALW294705-1KG-K
Propyl-3,4,5-trihydroxybenzoate, Sigma-Aldrich®
Propyl 3,4,5-trihydroxybenzoate
Formula: C₁₀H₁₂O₅
MW: 212.2 g/mol
Boiling Pt: 449
Melting Pt: 145…148 °C
Density: 1.21 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002196
CAS Number: 121-79-9
EINECS: 204-498-2
Merck Index: 12,08044