Isophthalic acid, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
I19209-2KG I19209-500G
SIALI19209-2KGEA 87.5 EUR
SIALI19209-2KG SIALI19209-500G
Isophthalic acid, Sigma-Aldrich®
Isophthalic acid
Formula: C₈H₆O₄
Boiling Pt: 119…121 °C (6 mmHg)
Melting Pt: 345…348 °C
Density: 1.54 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002516
CAS Number: 121-91-5
EINECS: 204-506-4
Merck Index: 13,05216