5,5'-Dithiobis(2-nitrobenzoic acid) (Ellmans reagent, DTNB), Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: DTNB, Ellman's reagent, 3,3'-Dithiobis(6-nitrobenzoic acid), 5,5'-Disulphanediylbis(2-nitrobenzoic acid), 2,2'-Dinitro-5,5'-dithiodibenzoic acid
M-Clarity™ quality level = MQ200
D8130-100G D8130-10G D8130-25G D8130-500MG D8130-5G
SIALD8130-100GEA 1980 EUR
SIALD8130-100G SIALD8130-10G SIALD8130-25G SIALD8130-500MG SIALD8130-5G
5,5'-Dithiobis(2-nitrobenzoic acid) (Ellmans reagent, DTNB), Sigma-Aldrich®
5,5'-Dithiobis(2-nitrobenzoic acid) (Ellmans reagent, DTNB)
Formula: C₁₄H₈N₂O₈S₂
MW: 396.36 g/mol
Melting Pt: 240…244 °C
Storage Temperature: Ambient
MDL Number: MFCD00007140
CAS Number: 69-78-3
EINECS: 200-714-4