2-Nitro-p-anisidine ≥99%
Supplier: Thermo Fisher Scientific
Danger
Synonyms: 4-Amino-3-nitroanisole, 4-Methoxy-2-nitroaniline
A11673.0E A11673.22 A11673.36
A11673.0EEA 464 EUR
A11673.0E A11673.22 A11673.36
2-Nitro-p-anisidine ≥99%
2-Nitro-p-anisidine
Purity: 99% CAS No.: 96-96-8 Molecular Formula: C7H8N2O3 Formula Weight: 168.15 Melting Point: 126-128° Beilstein Reference: 880318 UN No.: 2811 MDL No.: MFCD00007152
Formula: C₇H₈N₂O₃
MW: 168.15 g/mol
Boiling Pt: ∼250 °C (1013 hPa)
Melting Pt: 126…128 °C
Flash Pt: >200 °C
Storage Temperature: Ambient
MDL Number: MFCD00007152
CAS Number: 96-96-8
EINECS: 202-547-2
UN: 2811
ADR: 6.1 II