2-Nitro-p-anisidine

Supplier: Thermo Scientific
Danger

Synonyms: 4-Amino-3-nitroanisole, 4-Methoxy-2-nitroaniline

BTB07533.1 BTB07533.10 BTB07533.50
MAYBBTB07533.1EA 0 EUR
MAYBBTB07533.1 MAYBBTB07533.10 MAYBBTB07533.50
2-Nitro-p-anisidine
2-Nitro-p-anisidine
Formula: C₇H₈N₂O₃
MW: 168.15 g/mol
Boiling Pt: ∼250 °C (1013 hPa)
Melting Pt: 124…126 °C
Flash Pt: >200 °C
MDL Number: MFCD00007152
CAS Number: 96-96-8
EINECS: 202-547-2
UN: 2811
ADR: 6.1 II

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR