Benzeneselenenyl bromide, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Phenylselenyl bromide, Phenyl hypobromoselenoite, Bromoselenobenzene, Phenylselenenyl bromide
M-Clarity™ quality level = MQ100
243965-25G 243965-5G
SIAL243965-25GEA 238 EUR
SIAL243965-25G SIAL243965-5G
Benzeneselenenyl bromide, Sigma-Aldrich®
Benzeneselenenyl bromide
Formula: (C₆H₅)SeBr
MW: 235.97 g/mol
Boiling Pt: 229
Melting Pt: 57…60 °C
Density: 1.898 g/cm³ (65.5 °C)
MDL Number: MFCD00000047
CAS Number: 34837-55-3
EINECS: 252-238-1
UN: 2928
ADR: 6.1 II