4'-Hydroxy-3'-methoxyacetophenone, Sigma-Aldrich®
Supplier: Merck
Synonyms: 4-Hydroxy-3-methoxy-acetophenone, 1-(4-Hydroxy-3-methoxyphenyl)ethanone, Acetovanillone, Apocynin
M-Clarity™ quality level = MQ100
A10809-25G A10809-100G
SIALA10809-25GEA 32.8 EUR
SIALA10809-25G SIALA10809-100G
4'-Hydroxy-3'-methoxyacetophenone, Sigma-Aldrich®
4'-Hydroxy-3'-methoxyacetophenone
Formula: C₉H₁₀O₃
MW: 166.18 g/mol
Boiling Pt: 295
Melting Pt: 115
MDL Number: MFCD00008747
CAS Number: 498-02-2
EINECS: 207-854-5
Merck Index: 13,00746