1-Phenyl-1,2-propanedione, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: 1-Phenylpropane-1,2-dione

M-Clarity™ quality level = MQ200

223034-1G 223034-5G
SIAL223034-1GEA 57 EUR
SIAL223034-1G SIAL223034-5G
1-Phenyl-1,2-propanedione, Sigma-Aldrich®
1-Phenyl-1,2-propanedione
Formula: C₆H₅C(O)C(O)CH₃
Boiling Pt: 228 °C (1013 hPa)
Melting Pt: ? 20 °C
Density: 1.116 g/cm³ (20 °C)
MDL Number: MFCD00008755
CAS Number: 579-07-7
EINECS: 209-435-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1394d5392b739ebd2496078378d299949ea575ae&vid=b85d2a588e386bc41f5caca908cd1530a3da9131&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR