Cyclopropyl methyl ketone, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: 1-Cyclopropylethanone
M-Clarity™ quality level = MQ100
C120006-100G C120006-25G
SIALC120006-100GEA 120 EUR
SIALC120006-100G SIALC120006-25G
Cyclopropyl methyl ketone, Sigma-Aldrich®
Cyclopropyl methyl ketone
Formula: C₅H₈O
MW: 84.12 g/mol
Boiling Pt: 114
Melting Pt: –68 °C
Density: 0.899 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001297
CAS Number: 765-43-5
EINECS: 212-146-4
UN: 1992
ADR: 3 II