2,6,6-Trimethyl-2-cyclohexene-1,4-dione, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ300
W342106-100G-K W342106-1KG-K W342106-5KG-K
SIALW342106-100G-KEA 104 EUR
SIALW342106-100G-K SIALW342106-1KG-K SIALW342106-5KG-K
2,6,6-Trimethyl-2-cyclohexene-1,4-dione, Sigma-Aldrich®
2,6,6-Trimethyl-2-cyclohexene-1,4-dione
Formula: C₉H₁₂O₂
Boiling Pt: 214 °C (1013 hPa)
Melting Pt: 15 °C
Density: 1.03 g/cm³ (20 °C)
MDL Number: MFCD00043119
CAS Number: 1125-21-9
EINECS: 214-406-2