PIPES monosodium salt, Sigma-Aldrich®
Supplier: Merck
Synonyms: Piperazine-1,4-bis(2-ethanesulphonic acid) monosodium salt, Piperazine-N,N'-bis-2-ethanesulphonic acid monosodium salt, PIPES-Na, Sodium hydrogen piperazine-1,4-diethanesulphonate, 2,2'-(Piperazine-1,4-diyl)bis(ethanesulphonic) acid monosodium salt, Monosodium piperazine-1,4-diethanesulphonate
M-Clarity™ quality level = MQ200
P2949-100G P2949-25G P2949-500G
SIALP2949-100GEA 181 EUR
SIALP2949-100G SIALP2949-25G SIALP2949-500G
PIPES monosodium salt, Sigma-Aldrich®
PIPES monosodium salt
Formula: C₈H₁₇N₂NaO₆S₂
Storage Temperature: Ambient
MDL Number: MFCD00065472
CAS Number: 10010-67-0
EINECS: 233-005-3