1-(4-Fluorophenyl)piperazine 98%
Supplier: Thermo Scientific
|
Danger
|
AC18454.1
AC18454.10
AC18454.50
MAYBAC18454.1EA
0
EUR
MAYBAC18454.1
MAYBAC18454.10
MAYBAC18454.50
1-(4-Fluorophenyl)piperazine 98%
1-(4-Fluorophenyl)piperazine
Formula:
C₁₀H₁₃FN₂ Boiling Pt: 363 °C (1013 hPa) Melting Pt: 30…33 °C Flash Pt: >110 °C |
MDL Number:
MFCD00005960 CAS Number: 2252-63-3 EINECS: 218-846-6 UN: 3259 ADR: 8,III |
Specification Test Results
Appearance | Clear yellow Liquid after melting |
Infrared spectrum | Conforms |
Refractive index | 1.5585 to 1.5605 (20°C, 589 nm) |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...