Tungstic(VI) acid 99+%
Supplier: Thermo Fisher Scientific
Synonyms: Dihydrogen wolframate, Tungstic acid
207611000 207615000
ACRO207611000EA 67.6 EUR
ACRO207611000 ACRO207615000
Tungstic(VI) acid 99+%
Tungstic (VI) acid
Formula: H₂O₄W
MW: 249.85 g/mol
Density: 5.5 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011336
CAS Number: 7783-03-1
EINECS: 231-975-2
Order Now
Specification Test Results
Appearance Yellow to yellow-green Powder
Assay ≥99.0 %