Industrial gloves, HyFlex® 11-816
Supplier: Ansell
11-816/9 11-816/7 11-816/6 11-816/11 11-816/10 11-816/8
ANSE11-816/9EA 70.9 EUR
ANSE11-816/9 ANSE11-816/7 ANSE11-816/6 ANSE11-816/11 ANSE11-816/10 ANSE11-816/8
Industrial gloves, HyFlex® 11-816
Gloves
This nylon/spandex liner with foam nitrile coating provides unparalleled comfort and extreme tactility.
  • Ultra-thin ergonomic design provides extreme tactility to handle small parts easily
  • Silicone-free: No transfer of silicone contaminants to metal parts prior to painting
  • Skin-friendly: Proprietary washing process that efficiently extracts impurities to deliver a cleaner, more comfortable glove
  • FORTIX™ Abrasion Resistance Technology delivers extended wear time for enhanced worker safety and productivity
The combination between the thin liner and coating provide extreme comfort and a barehand feel for workers who rely on fingertip sensitivity for precision tasks. The FORTIX™ coating lasts longer than comparable styles under abrasive conditions for a longer wear life. HyFlex® 11-816 also ensures a secure grip in dry environments.
Recommended for: Picking, fastening components; assembling and mounting components; fastening components and parts; general handling; transport & logistics.
Certifications: EN 388:2016 (4121A); Cat. II
REACH Compliant
Packaging: 12 pairs per bag.