Potassium bromide, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ100
449962-10G 449962-50G
SIAL449962-10GEA 169 EUR
SIAL449962-10G SIAL449962-50G
Potassium bromide, Sigma-Aldrich®
Potassium bromide
Formula: KBr
Boiling Pt: 1435 °C (1013 hPa)
Melting Pt: 730 °C
Density: 2.75 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011358
CAS Number: 7758-02-3
EINECS: 231-830-3
Merck Index: 13,07701