Potassium bromide, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ200
451010-100G 451010-25G
SIAL451010-100GEA 298 EUR
SIAL451010-100G SIAL451010-25G
Potassium bromide, Sigma-Aldrich®
Potassium bromide
Formula: KBr
Boiling Pt: 1435 °C (1013 hPa)
Melting Pt: 730 °C
Density: 2.75 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011358
CAS Number: 7758-02-3
EINECS: 231-830-3
Merck Index: 13,07701