Rubidium chloride, Sigma-Aldrich®

Supplier: Merck

Synonyms: Rubidiumchloride

M-Clarity™ quality level = MQ200

204250-10G 204250-50G
SIAL204250-10GEA 281 EUR
SIAL204250-10G SIAL204250-50G
Rubidium chloride, Sigma-Aldrich®
Rubidium chloride
Formula: RbCl
MW: 120.92 g/mol
Boiling Pt: 1390
Melting Pt: 715
Density: 2.8 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011187
CAS Number: 7791-11-9
EINECS: 232-240-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR