Potassium sulphide (K2Sx), Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ100
12665-100G 12665-1KG 12665-6X1KG
SIAL12665-100GEA 40.7 EUR
SIAL12665-100G SIAL12665-1KG SIAL12665-6X1KG
Potassium sulphide (K2Sx), Sigma-Aldrich®
Potassium sulphide (K2Sx)
Formula: K₂(S)n
Melting Pt: 471 °C
Density: 1.65 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00148074
CAS Number: 37199-66-9
EINECS: 253-390-1
UN: 1382
ADR: 4.2,II