Calcium acetylide, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Calcium acetylide
21039-1KG-F 21039-100G-F
SIAL21039-1KG-FEA 133 EUR
SIAL21039-1KG-F SIAL21039-100G-F
Calcium acetylide, Sigma-Aldrich®
Calcium acetylide
Formula: CaC₂
MW: 64.1 g/mol
Melting Pt: 2300
Density: 2.22 g/cm³ (20 °C)
MDL Number: MFCD00010905
CAS Number: 75-20-7
EINECS: 200-848-3
UN: 1402
ADR: 4.3 II
Merck Index: 12,01696