Calcium acetylide, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Calcium acetylide
M-Clarity™ quality level = MQ100
270296-2KG 270296-500G
SIAL270296-2KGEA 202 EUR
SIAL270296-2KG SIAL270296-500G
Calcium acetylide, Sigma-Aldrich®
Calcium acetylide
Formula: CaC₂
MW: 64.1 g/mol
Melting Pt: 2300
Density: 2.22 g/cm³ (20 °C)
MDL Number: MFCD00010905
CAS Number: 75-20-7
EINECS: 200-848-3
UN: 1402
ADR: 4.3 II
Merck Index: 12,01696