Indium(I) iodide, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
578606-10G 578606-2G
SIAL578606-10GEA 666 EUR
SIAL578606-10G SIAL578606-2G
Indium(I) iodide, Sigma-Aldrich®
Indium (I) iodide
Formula: InI
MW: 241.72 g/mol
Boiling Pt: 711
Melting Pt: 351
Density: 5.32 g/cm³ (20 °C)
MDL Number: MFCD00135543
CAS Number: 13966-94-4
EINECS: 237-746-3
UN: 3260
ADR: 8 III