Sand (sea sand)
Supplier: Roth Carl
8441.1 8441.2
ROTH8441.1EA 0 EUR
ROTH8441.1 ROTH8441.2
Sand (sea sand)
Sea sand
Formula: SiO₂
MW: 60.08 g/mol
Boiling Pt: 2230
Melting Pt: 1710
Density: 2.66 g/cm³ (20 °C)
Storage Temperature: Ambient
CAS Number: 7631-86-9
EINECS: 231-545-4