1,4-Bis(diphenylphosphino)butane, Sigma-Aldrich®
Supplier: Merck
Synonyms: DPPB, 1,4-Bis(diphenylphosphino)butane, Butylenebis(diphenylphosphine), Bis(1,2-diphenylphosphino)butane, Butane-1,3-diylbis(diphenylphosphine)
M-Clarity™ quality level = MQ100
261947-25G 261947-5G
SIAL261947-25GEA 89.4 EUR
SIAL261947-25G SIAL261947-5G
1,4-Bis(diphenylphosphino)butane, Sigma-Aldrich®
1,4-Bis(diphenylphosphino)butane
Formula: C₂₈H₂₈P₂
MW: 426.48 g/mol
Melting Pt: 195
Density: 1.23 g/cm³ (20 °C)
MDL Number: MFCD00003051
CAS Number: 7688-25-7
EINECS: 231-698-7