Dodecyltrimethylammonium chloride, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: (1-Dodecyl)trimethylammonium chloride, N,N,N-Trimethyldodecan-1-aminium chloride, DTAC
M-Clarity™ quality level = MQ200
17104-100G 17104-25G
SIAL17104-100GEA 342 EUR
SIAL17104-100G SIAL17104-25G
Dodecyltrimethylammonium chloride, Sigma-Aldrich®
Dodecyltrimethylammonium chloride
Formula: C₁₅H₃₄ClN
Melting Pt: 248…249.5 °C
MDL Number: MFCD00041974
CAS Number: 112-00-5
EINECS: 203-927-0