Ferrocene, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
F408-100G
F408-500G
F408-5G
SIALF408-100GEA
43.4
EUR
SIALF408-100G
SIALF408-500G
SIALF408-5G
Ferrocene, Sigma-Aldrich®
Ferrocene
Formula:
C₁₀H₁₀Fe Boiling Pt: 249 °C (1013 hPa) Melting Pt: 172…175 °C Density: 1.49 g/cm³ (20 °C) |
MDL Number:
MFCD00001427 CAS Number: 102-54-5 EINECS: 203-039-3 UN: 1325 ADR: 4.1,II Merck Index: 13,04070 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...