Selenium dioxide, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ200
200107-25G 200107-5G
SIAL200107-25GEA 154 EUR
SIAL200107-25G SIAL200107-5G
Selenium dioxide, Sigma-Aldrich®
Selenium dioxide
Formula: SeO₂
MW: 110.96 g/mol
Melting Pt: 315
Density: 3.95 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003562
CAS Number: 7446-08-4
EINECS: 231-194-7
UN: 3283
ADR: 6.1 II
Merck Index: 13,08508