Cerium(IV) oxide, Sigma-Aldrich®

Supplier: Merck

Synonyms: Ceric oxide, Cerium dioxide

M-Clarity™ quality level = MQ200

211575-100G 211575-500G
SIAL211575-100GEA 70.5 EUR
SIAL211575-100G SIAL211575-500G
Cerium(IV) oxide, Sigma-Aldrich®
Cerium (IV) oxide
Formula: CeO₂
Boiling Pt: 3468 °C (1013 hPa)
Melting Pt: 2000 °C
Density: 7.13 g/cm³ (25 °C)
MDL Number: MFCD00010933
CAS Number: 1306-38-3
EINECS: 215-150-4
Merck Index: 12,02035

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR