Dansyl chloride, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: 5-(Dimethylamino)naphthalene-1-sulphonyl chloride
M-Clarity™ quality level = MQ200
D2625-1G D2625-25G
SIALD2625-1GEA 71.9 EUR
SIALD2625-1G SIALD2625-25G
Dansyl chloride, Sigma-Aldrich®
Dansyl chloride
Formula: C₁₂H₁₂ClNO₂S
MW: 269.75 g/mol
Melting Pt: 69…73 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00003985
CAS Number: 605-65-2
EINECS: 210-092-6
UN: 3261
ADR: 8 III
Merck Index: 13,02840