Silver hexafluoroantimonate ≥99%
Supplier: Thermo Fisher Scientific
Danger
Synonyms: Silver hexafluoroantimonate(V)
11538.03 11538.06 11538.14
11538.03EA 35.4 EUR
11538.03 11538.06 11538.14
Silver hexafluoroantimonate ≥99%
Silver hexafluoroantimonate
Purity: 99% CAS No.: 26042-64-8 Molecular Formula: AgSbF6 Formula Weight: 343.41 Physical Form: Powder/Lump Sensitivity: Light Sensitive and Hygroscopic Appearance: White to cream UN No.: 3260 MDL No.: MFCD00003401
Formula: AgF₆Sb
MW: 343.62
Density: 4.65…4.85 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003401
CAS Number: 26042-64-8
EINECS: 247-429-1
UN: 3260
ADR: 8 III