Titanium carbide, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
307807-100G
307807-500G
SIAL307807-100GEA
105
EUR
SIAL307807-100G
SIAL307807-500G
Titanium carbide, Sigma-Aldrich®
Titanium carbide
Formula:
CTi Boiling Pt: 4300 °C (1013 hPa) Melting Pt: 3067 °C Density: 4.93 g/cm³ (25 °C) |
MDL Number:
MFCD00011268 CAS Number: 12070-08-5 EINECS: 235-120-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...