Molybdenum dioxide, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
234761-10G 234761-50G
SIAL234761-10GEA 36 EUR
SIAL234761-10G SIAL234761-50G
Molybdenum dioxide, Sigma-Aldrich®
Molybdenum dioxide
Formula: MoO₂
MW: 127.94 g/mol
Boiling Pt: 1207…1307 °C (1013 hPa)
Melting Pt: 770…795 °C
Density: 6.47 g/cm³ (25 °C)
MDL Number: MFCD00014223
CAS Number: 18868-43-4
EINECS: 242-637-9