Iron(II) iodide, ultra dry ≥99.99% (metals basis)
Supplier: Thermo Fisher Scientific
Danger
Synonyms: Ferrous iodide, Iron diiodide
Ampuled under argon
13619.03 13619.09
13619.03EA 105 EUR
13619.03 13619.09
Iron(II) iodide, ultra dry ≥99.99% (metals basis)
Iron (II) iodide
Purity: 99.99% CAS No.: 7783-86-0 Molecular Formula: FeI2 Formula Weight: 309.66 Physical Form: -10 Mesh Beads, Ampouled under argon Density: 5.32 Appearance: Black Odour: Odourless UN No.: 3260 MDL No.: MFCD00016094
Formula: FeI₂
MW: 309.66 g/mol
Density: 5.32
Storage Temperature: Ambient
MDL Number: MFCD00016094
CAS Number: 7783-86-0
EINECS: 232-031-2
UN: 3260
ADR: 8 III
Merck Index: 12,04097