Palladium(II) acetylacetonate, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Palladium (II) 2,4-pentanedionate, Bis(2,4-pentanedionato-O,O')palladium (II), Palladium (II) bis(2,4-pentanedionato-O,O'), Palladium(II) 2,4-pentanedionate
M-Clarity™ quality level = MQ100
683388-2G 683388-500MG
SIAL683388-2GEA 134 EUR
SIAL683388-2G SIAL683388-500MG
Palladium(II) acetylacetonate, Sigma-Aldrich®
Palladium (II) acetylacetonate
Formula: C₁₀H₁₄O₄Pd
MW: 304.64 g/mol
Boiling Pt: ∼190 °C (1013 hPa)
Melting Pt: 190
MDL Number: MFCD00000025
CAS Number: 14024-61-4
EINECS: 237-859-8