Iron(III) acetylacetonate, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: Tris(2,4-pentanedionato-O,O')iron (III), Ferric acetylacetonate, Iron (III) 2,4-pentanedionate, Iron (III) tris(2,4-pentanedionato-O,O')

M-Clarity™ quality level = MQ200

44920-100G 44920-500G
SIAL44920-100GEA 65.4 EUR
SIAL44920-100G SIAL44920-500G
Iron(III) acetylacetonate, Sigma-Aldrich®
Iron (III) acetylacetonate
Formula: C₁₅H₂₁FeO₆
Boiling Pt: 100 °C (9.7513 mmHg)
Melting Pt: 180…183 °C
Density: 5.24 g/cm³ (20 °C)
MDL Number: MFCD00000020
CAS Number: 14024-18-1
EINECS: 237-853-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR