Methylene blue ≥82% (dye content), certified by the Biological Stain Commission, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Warning
Synonyms: Basic Blue 9, Methylthioninium chloride, Methylene blue chloride, 3,7-Bis(dimethylamino)phenothiazin-5-ium chloride, Swiss Blue
M-Clarity™ quality level = MQ200
M9140-100G M9140-25G M9140-100G M9140-25G
M9140-100GEA 291 EUR
M9140-100G M9140-25G SIALM9140-100G SIALM9140-25G
Methylene blue ≥82% (dye content), certified by the Biological Stain Commission, Sigma-Aldrich®
Methylene blue
Formula: C₁₆H₁₈ClN₃S
MW: 319.86 g/mol
Melting Pt: 180
Density: 1.757 g/cm³ (22 °C)
Storage Temperature: Ambient
MDL Number: MFCD00150008
CAS Number: 61-73-4
EINECS: 200-515-2
Merck Index: 12,06137
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=35017596fb0e86cf2655162ff7484bd23f024fd2&vid=473fa74160fa88b754101b22669323bc131effca&prid=web&clid=SAPDAM