Pyrazinamide, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ200

P7136-100G P7136-10G P7136-25G
SIALP7136-100GEA 301 EUR
SIALP7136-100G SIALP7136-10G SIALP7136-25G
Pyrazinamide, Sigma-Aldrich®
Pyrazinamide
Formula: C₅H₅N₃O
Boiling Pt: 357 °C (1013 hPa)
Melting Pt: 188…191 °C
MDL Number: MFCD00006132
CAS Number: 98-96-4
EINECS: 202-717-6
Merck Index: 12,08140

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR