Avantor® Hichrom HI-FFAP EXT, GC Columns
Supplier: Avantor
HI44-18-018-20EA
726
EUR
HI44-18-018-20
HI44-18-018-60
HI44-18-020-20
HI44-18-020-50
HI44-18-020-60
HI44-18-040-20
HI44-18-040-40
HI44-20-010-10
HI44-20-010-20
HI44-20-010-40
HI44-20-010-50
HI44-20-020-10
HI44-20-020-20
HI44-20-040-10
HI44-20-040-20
HI44-20-040-60
HI44-20-100-40
HI44-25-010-15
HI44-25-025-105
HI44-25-025-30
HI44-25-025-60
HI44-25-050-105
HI44-25-050-30
HI44-25-050-60
HI44-25-100-10
HI44-25-100-105
HI44-25-100-30
HI44-25-100-60
HI44-32-010-10
HI44-32-010-105
HI44-32-010-30
HI44-32-025-105
HI44-32-025-15
HI44-32-025-50
HI44-32-025-60
HI44-32-050-10
HI44-32-050-105
HI44-32-050-15
HI44-32-050-60
HI44-32-100-10
HI44-32-100-105
HI44-32-100-15
HI44-32-100-30
HI44-32-150-10
HI44-32-150-105
HI44-32-150-15
HI44-32-300-10
HI44-32-300-105
HI44-32-300-50
HI44-53-010-10
HI44-53-010-15
HI44-53-010-30
HI44-53-025-15
HI44-53-025-60
HI44-53-050-15
HI44-53-050-50
HI44-53-050-60
HI44-53-100-30
HI44-53-100-60
HI44-53-150-60
HI44-53-150-75
HI44-53-200-30
HI44-53-200-50
HI44-53-250-15
HI44-53-250-30
HI44-53-250-50
HI44-53-250-60
HI44-53-250-75
HI44-05-005-2
HI44-05-005-2.5
HI44-10-010-10
HI44-10-010-15
HI44-10-020-10
HI44-10-020-15
HI44-10-020-5
HI44-10-040-15
HI44-15-015-10
HI44-15-015-15
HI44-15-015-20
HI44-15-015-5
HI44-25-025-15G5
HI44-25-025-30G5
HI44-25-025-60G5
HI44-53-100-30G5
Avantor® Hichrom HI-FFAP EXT, GC Columns
Chromatography Columns
SPECIFICATIONS
Phase name | HI-FFAP EXT |
Environmentally Preferable | |
Phase composition | Acid modified Polyethylene Glycol (PEG) |
Polarity | Polar stationary phase |
Crossbond | Yes |
Temperature limit | 260 °C |
USP code | G14, G15, G16, G25, G35, G39 |